HPLC Safety Caps, V2.0, Preparative
HPLC Safety Caps, V2.0, Preparative
Actual product may vary
HPLC Safety Caps, V2.0, Preparative
SafetyCap is designed to safety extract solvents when working with any HPLC/UHPLC system. For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate. The SCAT Safety Caps provide maximum safety for your HPLC solvent supply.
Have Questions?
Catalog NumberSize (Metric)Includes
FIS3100-324.76 mm O.D. fitting3 fittings, 1 air valve
Catalog number FIS3100-32
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size (Metric):
4.76 mm O.D. fitting
Includes:
3 fittings, 1 air valve
  • Luer Lock Adapter
  • Improved fitting design for easier connection
  • Constructed of medical grade PTFE, PFA fittings, and a PPS screw cap
  • Includes space for individually labeling
  • Autocalvable up to 200°C
  • Stable construction for improved handling
Specifications
For Use With (Application)HPLC Systems
For Use With (Equipment)Lab Bottles
Includes3 fittings, 1 air valve
MaterialPFA, PTFE, PPS
Size (Metric)4.76 mm O.D. fitting
Thread SizeGL 45
TypeSafety Cap
Unit SizeEA