▸
Tubing
▸
Tubing Connectors and Fittings
▸
Reaction Tubes with High Vacuum Valve
Actual product may vary
Fisherbrand
Reaction Tubes with High Vacuum Valve
Cylindrical shaped tube used to connect two or more glass pieces
Have Questions?
Catalog Number
Capacity (Metric)
Cone Size
FIS31-501-426
100 mL
24/40
Catalog number
FIS31-501-426
Price (SGD)
210.00
Each
Add to cart
Capacity (Metric):
100 mL
Cone Size:
24/40
Price (SGD)
210.00
Each
Add to cart
With standard taper outer joint and side arm with high vacuum valve
Valves with PTFE O-ring at tip and PTFE wiper to protect Viton O-ring on shaft
Specifications
Description
100 mL, 0-4 mm
Autoclavable
Autoclavable
Material
Borosilicate Glass (3.3)
Cone Size
24/40
Color
Clear
Capacity (Metric)
100 mL
Product Type
Reaction Tube with High-vacuum Valve