MX35 Premier™ Disposable Low-profile Microtome Blades
Actual product may vary
MX35 Premier™ Disposable Low-profile Microtome Blades
MX35 Premier™ Disposable Low-profile Microtome Blades
MX35 Premier™ Disposable Low-profile Microtome Blades
MX35 Premier™ Disposable Low-profile Microtome Blades
Thermo Scientific
MX35 Premier™ Disposable Low-profile Microtome Blades
Section hard tissues and high-quality thin sections with high-performance with Epredia™ MX35 Premier™ disposable low-profile microtome blades.
Have Questions?
Catalog NumberProduct Type
FIS3051835Microtome Blade, Premier
Catalog number FIS3051835
Price (SGD)
-
Product Type:
Microtome Blade, Premier
  • Sharper edge with increased durability for longer blade life
  • Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life
Specifications
Product TypeMicrotome Blade, Premier
ProfileLow
For Use With (Application)Designed for routine histology and are good for a variety of tissue types, including hard tissue
DisposableSingle-use
Cutting Angle34°
Thickness (English)0.01 in.
Thickness (Metric)0.24 mm
Unit SizeCase