EP Reference Solution Y5, Yellow Reference Solution, Ricca Chemical
EP Reference Solution Y<sub>5</sub>, Yellow Reference Solution, Ricca Chemical
EP Reference Solution Y<sub>5</sub>, Yellow Reference Solution, Ricca Chemical
EP Reference Solution Y<sub>5</sub>, Yellow Reference Solution, Ricca Chemical
EP Reference Solution Y5, Yellow Reference Solution, Ricca Chemical
Have Questions?
Catalog NumberQuantity
FIS288354120 mL
Catalog number FIS288354
Price (SGD)
-
Quantity:
120 mL
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5, 7647-01-0, 7758-99-8, 10025-77-1, 7791-13-1, 7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialColor standard Y5
PackagingAmber Glass Bottle
CAS7732-18-5
CAS7647-01-0
CAS7758-99-8
View more