EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
Have Questions?
Catalog NumberQuantityPackaging
FIS2705-321 LNatural Poly Bottle
Catalog number FIS2705-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEDTA Titrant
ColorColorless
CAS6381-92-6
CAS Min %96.0
CAS Max %100.0
View more