▸
Personal Protective Equipment
▸
Safety Clothing
▸
Lab Coats and Jackets
▸
General Purpose Lab Coats and Jackets
▸
Extra-Safe™ Lab Coats — Cranberry
Actual product may vary
ValueMax
Extra-Safe™ Lab Coats — Cranberry
Static-free—no clinging
Have Questions?
Catalog Number
Size
FIS23-900-512A
Small
Catalog number
FIS23-900-512A
Price (SGD)
-
Size:
Small
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
Fluid-resistant, breathable
Snap front, three pockets, knit cuffs and collar
Knee length
Autoclavable and washable up to five times
Latex-free, fire retardant
Specifications
Product Type
General Purpose Lab Coat
Size
Small
Gender
Unisex
Material
SMS
Color
Cranberry
Closure Type
Snaps
Disposable
Single-use
Style
Knee-length
No. of Pockets
3
Latex Free
Yes
Thickness
3-ply
Certifications/Compliance
OSHA
Construction
3-ply SMS