Deionized Water
Thermo Scientific
Deionized Water
Thermo Scientific™ Deionized Water.
Have Questions?
Catalog NumberQuantity
FIS23-7516285 gal.
Catalog number FIS23-751628
Price (SGD)
-
Quantity:
5 gal.
Request bulk or custom format
Specifications
Quantity5 gal.
Unit SizeEA
Ozonated to inhibit microbial growth.
  • For use in flame photometry, blood gas humidifying chambers, electrode maintenance and instrument cleaning
Note:
Because of the many ways customers use this product, Thermo Fisher Scientific cannot provide an open container stability claim. Customers may establish their own open container stability claim by following CLSI guidelines.
Not intended for infusion, injection or irrigation.