Check Order Status
Water, deionized MilliporeSigma™
Element Search
Structure Search
Water, deionized MilliporeSigma™
Water; deionized; distilled; Type II; HARLECO; ; H
2
O
Have Questions?
Catalog Number
Quantity
Packaging
FIS23-749998
4 L
Poly Bottle
Catalog number
FIS23-749998
Price (SGD)
-
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Water
CAS
7732-18-5
Molecular Formula
H2O
Synonym
dihydrogen oxide, dihydrogen monoxide
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
View more