Specifications
Product TypeO.C.T. Compound
DescriptionCryostat Embedding Medium
MaterialPolyvinyl Alcohol, Polyethylene Glycol, Nonreactive Ingredients
ColorClear
Length (Metric)150 mm
Width (Metric)40 mm
Length (English)6 in.
Width (English)1.625 in.
Depth (Metric)40 mm
Depth (English)1.625 in.
For Use With (Application)Cryostat, Embedding Frozen Tissue Specimens
Quantity4 oz. (12/Case)
Unit SizeEach