Aquet™ Detergent
Aquet™ Detergent
Actual product may vary
Aquet™ Detergent
Aquet™ Detergent
Bel-Art
Aquet™ Detergent
For manual cleaning of glassware, plastics and surfaces
Have Questions?
Catalog NumberVolume (English)
FIS23-7210681 gal.
Catalog number FIS23-721068
Price (SGD)
-
Volume (English):
1 gal.
  • Phosphate-free, nonionic
  • Will not etch or mar precision glassware
  • Rinses quickly; dries without wiping
  • Each convenient, pre-portioned pouch makes one gallon of 1% solution
  • 3.8 liter (1 gallon) bottle comes with 473ml (16oz) working solution dispenser bottle
  • Not for use in automatic dishwashers
Specifications
Product TypeDetergent
Color-codedNo
FormLiquid
pH5 to 9
ScentRegular Scent
SterilityNon-sterile
Boiling Point>200°C
CompositionEthoxylated Nonylphenol, branched 19 to 21%, Dioxane <0.0006 %, Ethylene Oxide 0.0001%, Water, de
For Use With (Application)For hand washing Glass and Plastic labware
Size1 gal.
Volume (English)1 gal.
Volume (Metric)3.8 L
Unit SizeCS
Contents & Storage
15°C to 49°C