Bleach Solution Dispenser System
Actual product may vary
Bleach Solution Dispenser System
Eliminates waste associated with disinfecting laboratory surfaces with diluted bleach solutions
Have Questions?
Catalog NumberProduct Type
FIS23-640-500Bleach Dilution System
Catalog number FIS23-640-500
Price (SGD)
61.00
Each
Product Type:
Bleach Dilution System
Price (SGD)
61.00
Each
Unique, two-bottle trigger sprayer automatically mixes and dilutes 5.25% bleach with tap water to make a 10% bleach solution (5000ppm available chlorine).
  • Eliminates measuring, mixing, dating, and discarding bleach solution
  • Simply fill/refill water cartridge with tap water and lock in disposable bleach cartridge as needed
  • Sealed cartridge eliminates hazard of spilling bleach on skin and clothing
Bleach Dilution Sprayer
  • Ergonomic trigger sprayer facilitates high usage
  • Durable construction stands up to bleach
  • Includes sprayer head and water bottle
Bleach Solution Cartridge
  • 5.25% Sodium Hypochlorite Bleach
  • EPA-registered to kill Clostridium difficile spores in four minutes (EPA Reg. No. 75266-1 )
  • Each cartridge makes approximately 1 gallon of useable solution
  • Twelve 11oz. bleach cartridges
Specifications
Product TypeBleach Dilution System
Certifications/ComplianceEPA
Dilution5.25% Bleach/tap Water for 10:1 water-to-bleach solution
For Use With (Application)Eliminates waste associated with disinfecting laboratory surfaces with diluted bleach solutions
IncludesTwo bottle trigger sprayer head and water bottle
Test Time30 sec. to 4 min.
Unit SizeEach