Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Eye Protection and Face Protection
▸
Face Shields
▸
Critical Cover™ Coverall Face Shields
Actual product may vary
AlphaProTech
Critical Cover™ Coverall Face Shields
Good peripheral vision while acting as a shield against dangerous splashes in the work environment
Have Questions?
Change view
Catalog Number
Headband Material
Model
FIS23-309-829
Adjustable Hook-and-Loop-Closure Headband
Full-face
FIS23-309-828
Elastic Comfort Band
Full-face
2 Options
Catalog number
FIS23-309-829
Price (SGD)
815.00
Case
Add to cart
Headband Material:
Adjustable Hook-and-Loop-Closure Headband
Model:
Full-face
Price (SGD)
815.00
Case
Add to cart
Lightweight and easily fits over glasses
Double-sided anti-fog coating
Low distorting and 100% optically clear
Soft polyurethane foam headband fits snugly against the forehead
Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Specifications
Thickness (English)
7 mil shield
Lens Coating
Anti-fog
Description
Fits easily over glasses
Lens Tint
Clear
Headband Material
Adjustable Hook-and-Loop-Closure Headband
No. per Case
100/Cs.
Model
Full-face