▸
Personal Protective Equipment
▸
Eye Protection and Face Protection
▸
Face Shields
▸
Critical Cover™ Coverall Face Shields
Actual product may vary
AlphaProTech
Critical Cover™ Coverall Face Shields
Good peripheral vision while acting as a shield against dangerous splashes in the work environment
Have Questions?
Change view
Catalog Number
Headband Material
Model
FIS23-309-828
Elastic Comfort Band
Full-face
FIS23-309-829
Adjustable Hook-and-Loop-Closure Headband
Full-face
2 Options
Catalog number
FIS23-309-828
Price (SGD)
735.00
Case
Add to cart
Headband Material:
Elastic Comfort Band
Model:
Full-face
Price (SGD)
735.00
Case
Add to cart
Lightweight and easily fits over glasses
Double-sided anti-fog coating
Low distorting and 100% optically clear
Soft polyurethane foam headband fits snugly against the forehead
Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Specifications
Thickness (English)
7 mil shield
Lens Coating
Anti-fog
Description
Fits easily over glasses
Lens Tint
Clear
Headband Material
Elastic Comfort Band
No. per Case
100/Cs.
Model
Full-face