Cleanware™ Aqua-Clear™ Water Conditioner
Actual product may vary
Bel-Art
Cleanware™ Aqua-Clear™ Water Conditioner
Protects water baths and other water systems against the growth of algae, bacteria, molds and biofilms. Extends the useablity of water baths.
Have Questions?
Catalog number FIS23-278339
Price (SGD)
-
Protects water baths and other water systems against the growth of algae, bacteria, molds and biofilms. Extends the useablity of water baths.
  • Will not harm X-ray film when used in developing solutions
  • Protects pumps and thermostats
  • Prevents clogs from forming in tubing
  • Blue tint fades after 3 months as a reminder to change the water and add new conditioner
  • Treats up to 50 L of water
  • Does not contain any substance listed by California's Safe Drinking Water and Toxic Enforcement Act of 1986 (Proposition 65)
Specifications
Capacity (Metric)100 mL
Bacterial Content<0.1 CFU/mL
Daily Output1 to 10 L
Endotoxin Concentration<0.001 EU/mL
Feedwater SourcePotable Tap
Flow Rate1.1 L/min.
Product Water TypeAll
Total Organic Carbon<500 ppb
TypeWater Conditioner
For Use With (Equipment)Water bath
Unit SizeEach