Cannon™ Cold-Cranking Simulator Standards
Cannon™ Cold-Cranking Simulator Standards
Cannon™ Cold-Cranking Simulator Standards
Cannon
Cannon™ Cold-Cranking Simulator Standards
Synthetic base oil standards used to calibrate the Cannon Cold-Cranking Simulator (CCS) oil testing. Cannon CL Standards for Cold-Cranking Simulator provides certified dynamic viscosity data (in cP or mPa/s) at temperatures from -5° to -35°C.
Have Questions?
Catalog NumberViscosity
FIS23-023-7213500 cSt at -20°C
Catalog number FIS23-023-721
Price (SGD)
403.00
Each
Viscosity:
3500 cSt at -20°C
Request bulk or custom format
Price (SGD)
403.00
Each
Specifications
Recommended Storage23°F to 95°F (-5°C to +35°C)
Viscosity3500 cSt at -20°C
Physical FormLiquid
TechniqueCalibration
Certifications/ComplianceASTM D 5293, SAE, NIST
View more
  • Conform to SAE specification J300 and ASTM D 5293
  • Viscosities based on NIST value of 1.0016 centipoise for water at 20°C
  • Traceable to NIST
  • Supplied in one pint bottle