Check Order Status
Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical
65,750 ppm TDS as NaCl
Have Questions?
Catalog Number
Quantity
FIS2249-16
500 mL
Catalog number
FIS2249-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity/TDS Standard
Name Note
100mS/cm (100, 000μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
93.0
View more