Check Order Status
Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C (30,300 ppm TDS as NaCl), Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C (30,300 ppm TDS as NaCl), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS2248-16
500 mL
Natural Poly Bottle
Catalog number
FIS2248-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Conductivity/TDS Standard
Packaging
Natural Poly Bottle
CAS
7647-14-5
Concentration or Composition Notes
30,300ppm as NaCl
CAS
7732-18-5
View more