Check Order Status
Conductivity Standard, 1000 μS/cm at 25°C (495 ppm TDS as NaCl), Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 1000 μS/cm at 25°C (495 ppm TDS as NaCl), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS2243-16
500 mL
Natural Poly Bottle
Catalog number
FIS2243-16
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity/TDS Standard
Name Note
1mS/cm (1000μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
98.0
View more