Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Have Questions?
Catalog NumberQuantityPackaging
FIS223652-321 LNatural Poly Bottle
Catalog number FIS223652-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS7647-14-5, 7732-18-5
Specifications
Chemical Name or MaterialConductivity Standard
Name Note1.413mS/cm (1413 μmho/cm) at 25°C
PackagingNatural Poly Bottle
CAS7647-14-5
CAS Min %98.0
View more