Check Order Status
Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
Structure Search
Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS223602-16
500 mL
Catalog number
FIS223602-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Ricca Chemical Company Conductivity Standard
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
98.0
CAS Max %
100.0
View more