Check Order Status
Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS2236-16
500 mL
Natural Poly Bottle
Catalog number
FIS2236-16
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity Standard
Shelf Life
12 months
Name Note
10μS/cm (10μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7647-14-5
View more