Check Order Status
Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
Element Search
Structure Search
Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
1 mL = 59.5 mg CoCl
2
.
6H
2
O
Have Questions?
Catalog Number
Quantity
Packaging
FIS2210-16
500 mL
Amber Glass Bottle
Catalog number
FIS2210-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Cobaltous Chloride
Color
Red
Name Note
Red Primary Solution
CAS
7791-13-1
CAS
7647-01-0
View more