Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Miscellaneous Tools
▸
Mini Scraper
Actual product may vary
Dynamic Diagnostics
Mini Scraper
Help keep your laboratory environment clear of paraffin and hard-to-remove debris
Have Questions?
Catalog Number
Material
FIS22-899-265
Plastic
Catalog number
FIS22-899-265
Price (SGD)
78.00
Pack
Add to cart
Material:
Plastic
Price (SGD)
78.00
Pack
Add to cart
Ideal for cleaning around microtomes with its contoured design for easy gripping
Push button lock system allows for easy changing of the blade
Specifications
Product Type
Mini Scraper
Material
Plastic
For Use With (Application)
Contoured designed makes scraper easy to hold for easy cleaning around microtome's
Includes
Push button lock system