RotoFlex™ Tube Rotator
Actual product may vary
RotoFlex™ Tube Rotator
RotoFlex™ Tube Rotator
RotoFlex™ Tube Rotator
Adapt to different racks without tools to accommodate a wide range of tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSpeed Range
FIS22-505-00122 rpm
FIS22-505-00218 rpm
Catalog number FIS22-505-001
Price (SGD)
1,856.00
Each
Speed Range:
22 rpm
Price (SGD)
1,856.00
Each
  • Mix micro to centrifuge tubes-0.5 mL to 50 mL tubes- with these versatile rotators
  • Adjust the mixing angle of each rotisserie platform separately from 0 to 90°
  • Rotate at a fixed speed with the RotoFlex, or from 10 to 40 rpm with the RotoFlex Plus
  • Change the mixing direction to suit your application needs
These flexible tube rotators work with tubes sizes from 0.5 to 50 mL. The 360° rotation gently and thoroughly mixes samples. Additional features include an adjustable mixing angle from 0 to 90° for each of the 4 mixing panels independently. The RotoFlex system has a fixed speed or 22 rpm for the 120 V version or 18 rpm for the 240 V version, and the rotational direction is reversible by simply tapping the rotisserie in the direction required. The RotoFlex Plus system has a variable speed range of 10 to 40 rpm, with a 1 rpm resolution. The Plus system also has an oscillation mode that agitates tubes while rotating for additional sample mixing. Additional rotisseries are available for purchase separately.
Specifications
Oscillation360° Rotation
Rocking Angle0 to 90°
Rotating Angle0 to 90°
Speed Range22 rpm
Electrical Requirements120 V, 60 Hz
Voltage120 V
Temperature Range (Metric)4°C to 60°C
Rotation360°
Tube Capacity0.5 to 0.8 mL, 1.5 to 2 mL, 5 to 7 mL, 10 to 15 mL, 50 mL
Product TypeBenchtop Tube Rotator
Load Bearing Capacity (English)1.8 lb.
Certifications/ComplianceCE, RoHS/WEEE
Frequency60 Hz
Depth (English)6.06 in.
Depth (Metric)15.4 cm
Height (English)7.67 in.
Height (Metric)19.5 cm
Length (English)13.25 in.
Length (Metric)33.66 cm
Width (English)8.85 in.
Width (Metric)22.5 cm
Shipping Weight (English)4.06 lb.
Shipping Weight (Metric)4.06 kg
Weight (English)1.8 lb.
Weight (Metric)0.8 kg
ModelRotoFlex
For Use With (Application)Hybridization, DNA purification, sample mixing/resuspension, Protein binding, immunoprecipitation
For Use With (Equipment)Incubators and cold room settings
Humidity Range20 to 85%, Non-condensing
No. of Positions42 x 0.5 to 0.8mL/30 x 1.5 to 2mL/14 x 5 to 7mL/14 x 10 to 15mL/6 x 50mL
Unit SizeEach