Manual Key Differential Counter
Actual product may vary
LW Scientific
Manual Key Differential Counter
LW Scientific™ Manual Key Differential Counter are economical, durable, and will provide years of use in the demanding laboratory settings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Keys
FIS22-500-1318
NLG2250-01305
Catalog number FIS22-500-131
Price (SGD)
299.00
Each
No. of Keys:
8
Price (SGD)
299.00
Each
  • Traditionally differential counters have been manual gear and level machines
  • Offers both 5-key and 8-key manual counters for those traditional techs to count Baso, Eosin, Myelo, Juven, Stab, Seg, Lymph, and Mono cells
  • Feature pictures and names of each cell for easier identification
Specifications
Product TypeDifferential Cell Counter
No. of Keys8
StyleManual
Total Count999
Dimensions (L x W x H)10 x 3.75 x 1.75 in.
Weight (Metric)1 lb.
Frequency50/60 Hz
Voltage100 to 240 VAC
For Use With (Application)Counting
For Use With (Equipment)Microscopes
IncludesPower cord
Unit SizeEach