Large Weighted Safety Shield
Large Weighted Safety Shield
Actual product may vary
Bel-Art
Large Weighted Safety Shield
Transparent shield provides high impact resistance
Have Questions?
Catalog number FIS22-388421
Price (SGD)
2,088.00
EA
Add to cart
Price (SGD)
2,088.00
EA
Add to cart
Transparent shield provides high impact resistance
  • Laboratory workers are protected by a 4.7mm (0.188 in.) thick clear, polycarbonate barrier
  • Stable three-sided construction provides front and side protection against flying debris
  • Heavy steel perimeter base is coated with blue epoxy
  • Plated steel bolts, nuts and washers used for assembly
  • Outside base 19 1/2 in. (50cm) and 6 1/2 in. (17cm) front to back
  • Shields meet High Mass Impact Test and High Velocity Impact Test as defined by ANSI Z87.1, sections 9.11 and 9.12
Specifications
Height (English)30 in.
Height (Metric)76 cm
Lens TypeMeets ANSI High Mass Impact Test and High Velocity Impact Test
ShapeRectangular
Thickness (English)0.188 in.
Thickness (Metric)4.7 mm
Width (English)19.5 in.
Width (Metric)49.53 cm
Lens CoatingUncoated
Lens TintClear
Product TypeWeighted Safety Shield
Weight (English)18.5 lb.
Weight (Metric)8.4 kg
MaterialPolycarbonate
DisposableReusable
Lens MaterialPolycarbonate
Frame MaterialPolycarbonate
UV ProtectionNone
Unit SizeEA