Quaver™ and Quaverette™ Vibrating Spatulas
Actual product may vary
Bel-Art
Quaver™ and Quaverette™ Vibrating Spatulas
Accurately transfer small amounts of material
Have Questions?
Catalog NumberProduct TypeBlade MaterialShapeLength (English)
FIS22-385039Quaver™ Vibrating SpatulaStainless SteelFlat, Curved End7 in.
Catalog number FIS22-385039
Price (SGD)
266.00
Each
Product Type:
Quaver™ Vibrating Spatula
Blade Material:
Stainless Steel
Shape:
Flat, Curved End
Length (English):
7 in.
Price (SGD)
266.00
Each
  • Rotating the toothed thumb wheel vibrates the spatula, giving user control over transfer from spatula to weighing dish
  • Make accurate additions of precious materials as needed
  • Quaver™ spatula features a flat blade with curved end
Specifications
AutoclavableNot Autoclavable
ShapeFlat, Curved End
Product TypeQuaver™ Vibrating Spatula
For Use With (Equipment)Weighing dish or weighing boat or container
For Use With (Application)Accurately transfer small amounts of material, provides precise control of sample transfer from the blade to the weighing dish or weighing boat
Length (English)7 in.
Blade MaterialStainless Steel
IncludesAngled end, toothed thumb wheel
MaterialStainless Steel
Length (Metric)18 cm
Unit SizeEach