KIMBLE™ Disposable Large-Volume Pasteur Pipets
KIMBLE™ Disposable Large-Volume Pasteur Pipets
Actual product may vary
KIMBLE™ Disposable Large-Volume Pasteur Pipets
Economical flint glass pipet valuable for aliquoting samples for autoanalysis
Have Questions?
Catalog number FIS22-378893
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Single transfer handles 4mL
  • Designed for use with API Analytab System
Useful in all types of enteric diagnostic testing
Specifications
Certifications/ComplianceUSP type III
DescriptionBody Length: 8.89cm; 3.5 in.
MaterialSoda Lime Glass
SterilityNon-sterile
Length (English)5.75 in.
Length (English) Tip2.25 in.
Length (Metric) Tip5.71 cm
Volume (English)0.135 oz.
Volume (Metric)4 mL
Length (Metric)14.6 cm
TypeLarge Volume Pasteur Pipet
For Use With (Equipment)API Analytab System
Unit SizePK