Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring
Actual product may vary
Bel-Art
Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring
Adds turbulence relative to similar smooth size cylindrical bars. Manufactured in a registered ISO 9001:2008 facility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS22-36332576.2 mm12.7 mm3 in.0.5 in.
FIS22-331-42125.4 mm9.5 mm1 in.0.37 in.
FIS22-33144750.8 mm9.5 mm2 in.0.37 in.
Catalog number FIS22-363325
Price (SGD)
67.00
Each
Length (Metric):
76.2 mm
Diameter (Metric):
12.7 mm
Length (English):
3 in.
Diameter (English):
0.5 in.
Price (SGD)
67.00
Each
Adds turbulence relative to similar smooth size cylindrical bars. Manufactured in a registered ISO 9001:2008 facility.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility
  • Autoclave safe
Specifications
AutoclavableAutoclavable
Certifications/ComplianceFDA, USP Class VI, ISO 9001:2008
ColorWhite
Diameter (English)0.5 in.
Length (English)3 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)12.7 mm
Length (Metric)76.2 mm
MaterialAlnico Magnet
Quantity1
ShapePolygon
TypeMagnetic Stir Bar
Unit SizeEach