Specifications
Slide Capacity25
Closure TypeSnap-on
ColorBlack
For Use With (Equipment)Microscope Slide
IncludesNumbered slide partitions
Length (English) Slide3 in.
Length (Metric) Slide75 mm
MaterialPlastic
Width (English) Slide1 in.
Width (Metric) Slide25 mm
Unit SizePack