▸
Blood, Hematology and Coagulation Testing Products
▸
Blood Specimen Collection
▸
Microhematocrit Tubes
▸
Microhematocrit Capillary Tubes
Actual product may vary
Microhematocrit Capillary Tubes
Low-taper wall construction guarantees error less than 1% absolute, 0.4 hematocrit, relative
Have Questions?
Change view
Catalog Number
Closure Color
Additive
FIS22-362574
Blue
None
FIS22-362566
Red
Heparin
2 Options
Catalog number
FIS22-362574
Price (SGD)
17.00
Pack
Add to cart
Closure Color:
Blue
Additive:
None
Price (SGD)
17.00
Pack
Add to cart
Low-taper wall construction guarantees error less than 1% absolute, 0.4 hematocrit, relative
Ends can be sealed by sealing clay or flame
Manufactured from low-taper, soft glass
Red band on tube indicates heparinization; blue band indicates nonheparinized tube
Interior surface of heparinized tubes is evenly coated with ammonium heparin (4 to 7 USP units), which will not interfere with sodium determination
Taper does not exceed 0.0254mm in 75mm
Available plain or heparinized with not less than 2 USP units of ammonium heparinate
Color-coded tube tips and labels
Packaged in Fisher Scientific™ shaker-top vials for ease in dispensing
Specifications
Product Type
Microhematocrit Non-heparinized Capillary Tube
Additive
None
Material
Glass
Closure Type
Tube Sealant
Capacity (Metric)
70 μL
Diameter (Metric) Inner
0.11 to 0.12 cm
Closure Color
Blue
Length (Metric)
75 mm
Length (English)
2.9 in.
Diameter (English) Inner
0.043 to 0.047 in.