▸
Environmental Samplers
▸
General Purpose Sample Bags
▸
UV Guard™ UV Protection Bags
Actual product may vary
Minigrip
UV Guard™ UV Protection Bags
Prevent light-sensitive products from deterioration
Have Questions?
Catalog Number
Dimensions (L x W)
FIS22-311-215
12 x 12 in. (30.48 x 30.48cm)
Catalog number
FIS22-311-215
Price (SGD)
930.00
Case
Add to cart
Dimensions (L x W):
12 x 12 in. (30.48 x 30.48cm)
Price (SGD)
930.00
Case
Add to cart
Amber
With Color Line™ and premium zipper
Film contains UV blocker
Thickness of 3 mil
Available in different sizes
Specifications
Closure Type
Premium Zipper
Color
Amber
Material
Polyethylene
Thickness (English)
3 mil
Dimensions (L x W)
12 x 12 in. (30.48 x 30.48cm)
Length (English)
12 in.
Width (English)
12 in.
Product Type
UV Protection Bag