Check Order Status
Plain Glass Microscope Slides
Plain Glass Microscope Slides
Precleaned to be free of debris
Have Questions?
Catalog number
FIS22-310397
Price (SGD)
138.00
PK
Add to cart
Price (SGD)
138.00
PK
Add to cart
1mm thick slides manufactured from highest purity, corrosion-resistant clear sheet glass fully annealed to minimize chipping
Slides have improved surface wettability and are guaranteed nonsticking
Edges are precision ground for smoothness
Packed in environmental transport pack and hermetically sealed in a protective film to keep out moisture
Specifications
Frosted
1 End
Length (Metric)
76 mm
Material
Glass
Width (Metric)
25 mm
Length (English)
3 in.
Quantity
1440/Cs.
Width (English)
1 in.
Dimensions (L x W)
3 x 1 in. (75 x 25 mm)