Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper
Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper
Actual product may vary
DWK Life Sciences
Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper
Ideal for shelf storage
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-308622250 mL
Catalog number FIS22-308622
Price (SGD)
1,370.00
CS
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
1,370.00
CS
Add to cart
  • Amber borosilicate glass bottle with narrow mouth
  • Protects light-sensitive reagents and solutions
  • Ground-glass octagon flat stopper provides excellent seal; protects top lip of bottle from dust
Specifications
Capacity8.45 oz., 250mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorAmber
Diameter (English)2.83 in.
MaterialGlass
Closure ColorAmber
MouthNarrow
Height (Metric)145 mm
Height (English)5.7 in.
Closure MaterialBorosilicate Glass
Diameter (Metric)72 mm
Unit SizeCS