Lab Guard™ Specimen Bags
Actual product may vary
Minigrip
Lab Guard™ Specimen Bags
Protect from inadvertent exposure to leakage and contamination
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)Product TypeQuantity
FIS22-306-3256 x 4 in. (15.2 x 10.1cm)1.75 milBiohazard Specimen Bag1000/Cs.
FIS22-3062979 x 6 in. (23 x 15cm)1.75 milSpecimen Transport Bag1000/Dispenser Bag
Catalog number FIS22-306-325
Price (SGD)
166.00
Case
Dimensions (L x W):
6 x 4 in. (15.2 x 10.1cm)
Thickness (English):
1.75 mil
Product Type:
Biohazard Specimen Bag
Quantity:
1000/Cs.
Price (SGD)
166.00
Case
Bags with the destroyable biohazard symbol feature allow uncontaminated bag remains to be disposed in regular waste; if bag remains are contaminated, dispose in RED waste.
  • Zippered polyethylene three-wall bags with attached document compartment
  • Sample and paperwork stay together to reduce clerical errors and misidentification
  • TearZone™ EZ open feature means no leaks
  • Available with convenient DispenserBag™ and DispenserBox packaging
Specifications
Description3-Walled Biohazard Specimen Bag
Closure TypeZip Seal
For Use With (Application)For transportation of specimens
MaterialPolyethylene
Quantity1000/Cs.
Thickness (English)1.75 mil
Dimensions (L x W)6 x 4 in. (15.2 x 10.1cm)
Length (English)6 in.
Width (English)4 in.
Product TypeBiohazard Specimen Bag
Length (Metric)15.2 cm
Width (Metric)10.1 cm
Unit SizeCase