Clear Plastic Wrap
Clear Plastic Wrap
Actual product may vary
Clear Plastic Wrap
Clear Plastic Wrap is designed specifically for the laboratory.
Have Questions?
Catalog NumberLength (English)
FIS22-305654100 ft.
Catalog number FIS22-305654
Price (SGD)
13.35
Special offer
Ends: 15-Dec-2025
35.00
Save 21.65 (62%)
EA
Add to cart
Length (English):
100 ft.
Price (SGD)
13.35
Special offer
Ends: 15-Dec-2025
35.00
Save 21.65 (62%)
EA
Add to cart
Clear Plastic Wrap is designed specifically for the laboratory.
  • Made of 100% ultraclean polyethylene—ensures sample purity and eliminates contamination
  • High-cling properties make it ideal for wrapping of beakers, flasks, or Petri dishes with an extraordinarily tight seal that stays in place
  • Low-tangle properties ensure wrap won't ball up when being handled
  • Carton has convenient metal tear-off bar
Specifications
ApplicationsLaboratory use
Length (English)100 ft.
ColorClear
Product TypeWrapping Film
Width (English)12 in.
Width (Metric)30 cm
Length (Metric)30.4 m
MaterialPolyethylene
For Use With (Equipment)Sealing, Weighing, Transferring Samples
Unit SizeEA