Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Flow Analysis
▸
Anemometer and Flowmeter Accessories
▸
Specialty Chemical Snoop™ Liquid Leak Detector
Actual product may vary
Ohio Valley Specialty Chemical
Specialty Chemical Snoop™ Liquid Leak Detector
Used for locating leaks in air and carrier gas lines
Have Questions?
Catalog Number
Quantity
FIS22-289334
8 oz.
Catalog number
FIS22-289334
Price (SGD)
59.00
Each
Add to cart
Quantity:
8 oz.
Price (SGD)
59.00
Each
Add to cart
236mL (8 oz.) squeeze bottle with 30.5cm (12 in.) snooper tube
Nontoxic, nonflammable and noncorrosive
Temperature range: -2 to +93°C (27 to 200°F)
For external surface use only
Specifications
Type
Leak Detector
For Use With (Application)
Locating leaks in air and carrier gas lines
Max. Operating Temperature (English)
200°F
Max. Operating Temperature (Metric)
93°C
Quantity
8 oz.