Viscosity Standards
Cannon
Viscosity Standards
Provide kinematic viscosities (in cSt or mm2/s), densities (in g/mL), and viscosities (incP or mPa/s) at many temperatures, ranging from -55° to +150°C. Cannon Certified Viscosity Standards are prepared in Cannon's ISO 9001:2000-registered and A2LA-accredited laboratory.
Have Questions?
Change viewbuttonViewtableView
Viscosity:
1.2 cSt at 100°C
3.0 cSt at 100°C
20 cSt at 100°C
120 cSt at 100°C
Catalog number FIS22-288554
Price (SGD)
337.00
Each
Viscosity:
1.2 cSt at 100°C
Request bulk or custom format
Price (SGD)
337.00
Each
Specifications
Recommended Storage-67°F to +302°F (-55°C to +150°C)
Viscosity1.2 cSt at 100°C
Physical FormLiquid
TechniqueCalibration and Verification
Certifications/ComplianceASTM D 2162, ISO/IEC 17025, NIST
View more
Provide kinematic viscosities (in cSt or mm2/s), densities (in g/mL), and viscosities (incP or mPa/s) at many temperatures, ranging from -55° to +150°C. Cannon Certified Viscosity Standards are prepared in Cannon's ISO 9001:2000-registered and A2LA-accredited laboratory.
  • Values for viscosity, kinematic viscosity, and density at a variety of temperatures are printed on the container label
  • Clearly identify your unit of viscosity measure (centistokes or centipoises) when ordering
  • Custom-blended and/or added temperature standards can be provided—additional charges will apply