No-Wire™ Autoclavable Test Tube Racks
No-Wire™ Autoclavable Test Tube Racks
Actual product may vary
Bel-Art
No-Wire™ Autoclavable Test Tube Racks
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositions
FIS22-288145Blue21
FIS22-269954Blue60
FIS22-269979White40
FIS22-269948White60
FIS22-288141White21
FKS12652966Blue21
FKS12662966Red40
Catalog number FIS22-288145
Price (SGD)
66.00
Each
Add to cart
Color:
Blue
Positions:
21
Price (SGD)
66.00
Each
Add to cart
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
  • Suitable for use inside and outside the water bath to meet the demands of daily lab use
  • Heavier than water so the rack remains stable when submerged, even when agitated
  • Store flat to save space, but assembles quickly and easily
  • Features grids with numbered and lettered rows for tube identification
  • Interlocking feet facilitate easy stacking
  • Sturdy polypropylene is steam autoclavable at 121°C (250°F)
  • Round racks fit into 1000mL beaker and include removable, snap-on handle for easy retrieval
  • Rectangular racks measure 24.6 x 10.4 x 6.4cm (9.7 x 4.1 x 2.5 in.) and feature end plates for labeling
Specifications
AutoclavableAutoclavable
ColorBlue
HoldsTubes (25 to 30 mm)
MaterialPolypropylene
Positions21
ShapeRectangular
StackableYes
SterilityNon-sterile
Dimensions (L x W x H)9.68 x 4.09 x 2.51 in. (24.59 x 10.4 x 6.4 cm)
Length (Metric)24.59 cm
Length (English)9.68 in.
Width (English)4.09 in.
Height (English)2.51 in.
Diameter (Metric) Opening30 mm
Product TypeTube Rack
Diameter (Metric) Inner30 mm
Shelves3 Tiers
Height (Metric)6.4 cm
Array3 x 7
Width (Metric)10.4 cm
Unit SizeEach