Microbank™ Bacterial and Fungal Preservation System
Actual product may vary
Microbank™ Bacterial and Fungal Preservation System
Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
Have Questions?
Catalog NumberClosure Color
FIS22-286-153Blue
Catalog number FIS22-286-153
Price (SGD)
-
Closure Color:
Blue
Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)
Specifications
Product TypeCryogenic Storage Vial
Closure ColorBlue
For Use With (Application)Designed to simplify the storage and retrieval of bacterial and fungal cultures
Quantity80 Pk.
Storage Requirements-70°C
Unit SizePack