Swab-Pak™ Swabs
Swab-Pak™ Swabs
Actual product may vary
Swab-Pak™ Swabs
Polyester-tipped, general-use swabs
Have Questions?
Catalog NumberTip TypeHandle Material
FIS22-281659Dual EndPlastic
Catalog number FIS22-281659
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Tip Type:
Dual End
Handle Material:
Plastic
Swabs are contained in a sterile, plastic transport tube with a tamper-evident label
Single swabs have a straight wire shaft on a plastic handle
  • Dual swabs have a plastic shaft
Specifications
For Use With (Application)General-use swabs
IncludesPlastic Tamper-evident labeled transport tube
SterilitySterile
Handle MaterialPlastic
Product TypeSwab
Tip MaterialDacron
Tip TypeDual End
Unit SizePK