Spinpak™ Magnetic Stirring Bars
Spinpak™ Magnetic Stirring Bars
Actual product may vary
Bel-Art
Spinpak™ Magnetic Stirring Bars
Provide instant availability of highly popular octagon shaped bars
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Length (English)
FIS22-27908425.4 mm1 in.
FIS22-27908312.7 mm0.5 in.
Catalog number FIS22-279084
Price (SGD)
81.00
PK
Add to cart
Length (Metric):
25.4 mm
Length (English):
1 in.
Price (SGD)
81.00
PK
Add to cart
  • Interrupted profile of octagonal shape provides more surface area and greater turbulence than comparable cylindrical stirring bars
  • Made from Teflon™ PTFE
  • ALNICO V encapsulated magnet
  • Autoclavable
Teflon™ is a registered trademark of the Chemours Company FC LLC used under license by Bel-Arts Products, Inc.
Specifications
Certifications/ComplianceFDA
Diameter (English)0.31 in.
Length (English)1 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)8 mm
Length (Metric)25.4 mm
MaterialAlnico Magnet
Quantity20
ShapeOctagon
TypeMagnetic Stir Bar Set
Unit SizePK