Fenske Routine Viscometers, CFRC Series
Actual product may vary
Cannon
Fenske Routine Viscometers, CFRC Series
For measuring kinematic viscosity of transparent Newtonian liquids, particularly petroleum products or lubricants, according to ASTM D 445 and ISO 3104.
Have Questions?
Catalog NumberViscosity RangeSize
FIS22-2739189000 to 45,000 cSt650
Catalog number FIS22-273918
Price (SGD)
564.00
Each
Viscosity Range:
9000 to 45,000 cSt
Size:
650
Price (SGD)
564.00
Each
  • Glass capillary
  • Available in Calibrated and Uncalibrated versions
  • Size: 25 to 700mm
  • Minimum sample volume: 7mL
Specifications
Product TypeRoutine Viscometer
Viscosity Range9000 to 45,000 cSt
CalibratedYes
Approximate Constant100mm2/s2
Certifications/ComplianceASTM D446, ISO 3105
IncludesCertificate of calibration and instruction sheet
Accuracy±0.2%
For Use With (Application)Measuring kinematic viscosity of transparent newtonian liquids, particularly petroleum products or lubricant
Size650
Volume (Metric)7 mL
Unit SizeEach