Octagon Spinbar™ Magnetic Stirring Bars
Octagon Spinbar™ Magnetic Stirring Bars
Actual product may vary
Bel-Art
Octagon Spinbar™ Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape for stirring small volumes in vessels such as vials, tubes and gradient makers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS22-27183338 mm9.5 mm1.5 in.0.37 in.
FIS22-06691022 mm8 mm0.87 in.0.31 in.
FIS22-06696928.6 mm8 mm1.12 in.0.31 in.
FIS22-06715722 mm9.5 mm0.87 in.0.37 in.
FIS22-06718163.5 mm9.5 mm2.5 in.0.37 in.
Catalog number FIS22-271833
Price (SGD)
23.00
Each
Add to cart
Length (Metric):
38 mm
Diameter (Metric):
9.5 mm
Length (English):
1.5 in.
Diameter (English):
0.37 in.
Price (SGD)
23.00
Each
Add to cart
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Specifications
AutoclavableAutoclavable
Certifications/ComplianceFDA, USP Class VI, ISO 9001:2008
ColorWhite
Diameter (English)0.37 in.
Length (English)1.5 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)9.5 mm
Length (Metric)38 mm
MaterialAlnico Magnet
QuantityEach
ShapeOctagon
TypeMagnetic Stir Bar with Pivot Ring
Unit SizeEach