Premium Microscope Slide Box, 100-Place, Cork, Black
Premium Microscope Slide Box, 100-Place, Cork, Black
Actual product may vary
Premium Microscope Slide Box, 100-Place, Cork, Black
Premium Microscope Slide Box, 100-Place, Cork, Black
Premium Microscope Slide Box, 100-Place, Cork, Black
100-Slide capacity
Have Questions?
Catalog number FIS22-267294
Price (SGD)
41.00
EA
Add to cart
Price (SGD)
41.00
EA
Add to cart
  • Molded, rigid boxes securely hold slides in grooved slots
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Box is lined with cork
  • Lid closes securely with a rust-resistant, nickel-plated, clasp and pin hinge
  • Stackable
Specifications
Slide Capacity100
Closure TypeHinged
ColorBlack
Depth (English)6.75 in.
Depth (Metric)17.1 cm
Device HoldsSlides
For Use With (Equipment)Microscope Slide
Height (English)1.3 in.
Height (Metric)3.3 cm
IncludesNumbered slots
Length (English)8.18 in.
Length (English) Slide3 in.
Length (Metric)208 mm
Length (Metric) Slide25 x 75 mm
MaterialPolystyrene
Width (English)8.75 in.
Width (English) Slide1 in.
Width (Metric)22.22 cm
Width (Metric) Slide25 x 75 mm
Unit SizeEA