Check Order Status
Extra-Thick Microscope Slides
Actual product may vary
Extra-Thick Microscope Slides
Approximately 20% thicker than standard slides
Have Questions?
Catalog Number
Width (English)
FIS22-267013
2 in.
Catalog number
FIS22-267013
Price (SGD)
261.00
PK
Add to cart
Width (English):
2 in.
Price (SGD)
261.00
PK
Add to cart
Approximately 20% thicker than standard slides
Made of clear, corrosion-resistant glass
Precleaned, with flat surfaces and ground edges for consistent quality
Frosted-end slides have sandblasted area on one end for positive sample identification
Specifications
Edges
Ground
Frosted
Yes
Length (Metric)
76.2 mm
Material
Glass
Precleaned
Yes
Thickness (Metric)
1.2 mm
Type
Microscope Slide
Width (Metric)
50.8 mm
Color
Clear
Length (English)
3 in.
Quantity
144/Pk.
Width (English)
2 in.