Cover Glasses: Rectangles
Cover Glasses: Rectangles
Actual product may vary
Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)Product Type
FIS22-2669080.17 to 0.25 mm22 mm30 mmCoverslip
FIS12-543A0.17 to 0.25 mm40 mm22 mmCoverslip
FIS12-543C0.17 to 0.25 mm50 mm22 mmCoverslip
FIS12-543F0.17 to 0.25 mm50 mm45 mmCoverslip
FIS12-544D#1.5 (0.16 to 0.19 mm)50 mm22 mmCoverslip
FIS12-544E#1.5 (0.16 to 0.19 mm)50 mm24 mmCoverslip
FIS12-544G#1.5 (0.16 to 0.19 mm)60 mm22 mmCoverslip
FIS12-544A#1.5 (0.16 to 0.19 mm)30 mm22 mmCoverslip
FIS12-544B#1.5 (0.16 to 0.19 mm)40 mm22 mmCoverslip
FIS12-544C#1.5 (0.16 to 0.19 mm)40 mm24 mmCoverslip
FIS12-545C#1 (0.13 to 0.16 mm)40 mm22 mmCoverslip
FIS12-545E#1 (0.13 to 0.16 mm)50 mm22 mmCoverslip
FIS12-545H#1 (0.13 to 0.16 mm)50 mm45 mmCoverslip
FIS12-545J#1 (0.13 to 0.16 mm)60 mm22 mmCoverslip
Catalog number FIS22-266908
Price (SGD)
-
Thickness (Metric):
0.17 to 0.25 mm
Length (Metric):
22 mm
Width (Metric):
30 mm
Product Type:
Coverslip
Made of finest optical borosilicate glass, with uniform thickness and size
Rectangular shape.
  • Corrosion-resistant
  • Lint-free and ready to use
Specifications
Product TypeCoverslip
ShapeRectangle
MaterialBorosilicate Glass
Length (Metric)22 mm
Width (Metric)30 mm
Thickness (Metric)0.17 to 0.25 mm
For Use With (Application)Microscope, histology, cytology, hematology, microbiology
Unit SizePK