CytoSoft™ Cytology Brush
CytoSoft™ Cytology Brush
Actual product may vary
CytoSoft™ Cytology Brush
Small, smooth brush fibers create a monolayer of cells that enhance interpretation
Have Questions?
Catalog NumberProduct TypeSterility
FIS22-263358Cytology BrushNon-sterile
Catalog number FIS22-263358
Price (SGD)
755.00
CS
Add to cart
Product Type:
Cytology Brush
Sterility:
Non-sterile
Price (SGD)
755.00
CS
Add to cart
  • The CytoSoft nylon brush increases collection of endocervical cells and component material
  • Increased cellular yield reduces the liability associated with inadequate specimens and false negative specimens obscured by blood during sampling
Specifications
Product TypeCytology Brush
MaterialNylon
SterilityNon-sterile
Length (English)8 in.
Length (Metric)20.3 cm
For Use With (Application)Used to collect specimens for sexually transmitted disease (STD) testing
Quantity500 Cs.
Unit SizeCS