Wheaton™ 90-Position Vial Rack
Wheaton™ 90-Position Vial Rack
Actual product may vary
DWK Life Sciences
Wheaton™ 90-Position Vial Rack
Save valuable lab space with this stackable and autoclavable vial rack
Have Questions?
Catalog NumberPositions
FIS22-26295590
Catalog number FIS22-262955
Price (SGD)
710.00
CS
Add to cart
Positions:
90
Price (SGD)
710.00
CS
Add to cart
Save valuable lab space with this stackable and autoclavable vial rack
  • Polypropylene construction
  • 6 wells deep x 15 wells wide
  • Alpha-numeric indexing
  • Bottom drainage
  • 17.1 mm well diameter; L x W x H: 33 x 17 x 3 cm (13 x 6.7 x 1.2 in.);
Specifications
Product TypeVial Rack
Array4 x 12
AutoclavableAutoclavable
ColorWhite
MaterialPolypropylene
Positions90
StackableYes
For Use With (Equipment)50 Cryule 1 and 2mL 12mm dia. round bottom autosampler vials with screw caps or 12 x 75mm Glass culture tubes
Dimensions (L x W x H)13 x 6.6 x 1.25 in. (33 x 17 x 3cm)
Length (Metric)33 cm
Length (English)13 in.
Width (Metric)17 cm
Width (English)6.6 in.
Height (Metric)3 cm
Height (English)1.25 in.
Diameter (Metric) Well17.1 mm
Diameter (Metric) Inner17.1 mm
Unit SizeCS