SP Scienceware™ Slide Staining and Storage Apparatus
Actual product may vary
Bel-Art
SP Scienceware™ Slide Staining and Storage Apparatus
Conveniently stain and store gels
Have Questions?
Catalog NumberProduct Type
FIS22-261920Slide Rack
Catalog number FIS22-261920
Price (SGD)
-
Product Type:
Slide Rack
  • Versatile slide rack and vessel for radiography gel staining and staining protocols
  • PMPA plastic construction makes the unit virtually shatterproof, lightweight, with exceptional solvent and heat resistance for extended use in cytology and histology labs
  • The rack and dish can simply be cleaned and are highly resistant to standard stains used in staining protocols
Specifications
Product TypeSlide Rack
Slide Capacity25
ColorBlack
Reagent Capacity250 mL
For Use With (Application)Used in cytology and histology labs
For Use With (Equipment)Slide Stainer
Temperature Range (Metric)90°C
Height (English)0.78 in.
Height (Metric)2 cm
Length (English)3.6 in.
Length (Metric)9 cm
Width (English)3 in.
Width (Metric)7.8 cm
Voltage0/1 V
Unit SizeEach